accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3160-37-0

IUPAC Name: (3E)-4-(2H-1,3-benzodioxol-5-yl)but-3-en-2-one
Synonyms: (3E)-4-(2H-1,3-benzodioxol-5-yl)but-3-en-2-one
Molecular Weight (g/mol): 190.2
Molecular Formula: C11H10O3
InChi Key: XIYPXOFSURQTTJ-NSCUHMNNSA-N
SMILES: CC(=O)\C=C\C1=CC=C2OCOC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results