accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 31608-22-7

IUPAC Name: 2-(4-bromobutoxy)oxane
Synonyms: 2-(4-bromobutoxy)oxane
Molecular Weight (g/mol): 237.14
Molecular Formula: C9H17BrO2
InChi Key: DFXFQWZHKKEJDM-UHFFFAOYNA-N
SMILES: BrCCCCOC1CCCCO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results