accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3162-29-6

IUPAC Name: 1-(2H-1,3-benzodioxol-5-yl)ethan-1-one
Synonyms: 1-(2H-1,3-benzodioxol-5-yl)ethanone
Molecular Weight (g/mol): 164.16
Molecular Formula: C9H8O3
InChi Key: BMHMKWXYXFBWMI-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C2OCOC2=C1
Percent Purity 
  • (1)
  • (7)
Quantity 
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (3)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (1)
  • (5)
  • (1)
  • (7)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (3)