accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COONOO

CAS RN 31680-08-7

IUPAC Name: 4-methoxy-3-nitrobenzaldehyde
Synonyms: 4-methoxy-3-nitrobenzaldehyde
Molecular Weight (g/mol): 181.15
Molecular Formula: C8H7NO4
InChi Key: YTCRQCGRYCKYNO-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=O)C=C1[N+]([O-])=O
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)