accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3OONHNNOOCH3H3CCH3OHOHN

CAS RN 31687-58-8

IUPAC Name: 3-{1-[(tert-butoxy)carbonyl]-1H-imidazol-4-yl}-2-{[(tert-butoxy)carbonyl]amino}propanoic acid; N-cyclohexylcyclohexanamine
Synonyms: 2-[(tert-butoxycarbonyl)amino]-3-[1-(tert-butoxycarbonyl)imidazol-4-yl]propanoic acid; dicha
Molecular Weight (g/mol): 536.71
Molecular Formula: C28H48N4O6
InChi Key: WBGMQHNUPJENDC-UHFFFAOYNA-N
SMILES: C1CCC(CC1)NC1CCCCC1.CC(C)(C)OC(=O)NC(CC1=CN(C=N1)C(=O)OC(C)(C)C)C(O)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)