accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 317-20-4

IUPAC Name: 7-fluoro-2,3-dihydro-1H-indole-2,3-dione
Synonyms: 7-fluoro-1H-indole-2,3-dione
Molecular Weight (g/mol): 165.12
Molecular Formula: C8H4FNO2
InChi Key: HGBGVEOXPHGSOS-UHFFFAOYSA-N
SMILES: FC1=CC=CC2=C1NC(=O)C2=O
Molecular Weight (g/mol) 
  • (1)
  • (5)
Percent Purity 
  • (2)
  • (1)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (5)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)