accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFFFFSNH2

CAS RN 317319-15-6

IUPAC Name: 3,5-bis(trifluoromethyl)benzene-1-carbothioamide
Synonyms: 3,5-bis(trifluoromethyl)benzenecarbothioamide
Molecular Weight (g/mol): 273.2
Molecular Formula: C9H5F6NS
InChi Key: OWYYZIVORPIBPZ-UHFFFAOYSA-N
SMILES: NC(=S)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results