accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOHNO

CAS RN 317854-65-2

IUPAC Name: 3-formyl-1H-indole-7-carboxylic acid
Synonyms: 3-formyl-1H-indole-7-carboxylic acid
Molecular Weight (g/mol): 189.17
Molecular Formula: C10H7NO3
InChi Key: HABYJDPNVBUDGD-UHFFFAOYSA-N
SMILES: OC(=O)C1=C2NC=C(C=O)C2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results