accessibility menu, dialog, popup

UserName

Viewing profile as user:
SNHNHN

CAS RN 3179-31-5

IUPAC Name: 2,3-dihydro-1H-1,2,4-triazole-3-thione
Synonyms: 1,2-dihydro-1,2,4-triazole-3-thione
Molecular Weight (g/mol): 101.13
Molecular Formula: C2H3N3S
InChi Key: AFBBKYQYNPNMAT-UHFFFAOYSA-N
SMILES: S=C1NNC=N1
Percent Purity 
  • (1)
  • (3)
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (4)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)