accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3179-76-8

IUPAC Name: (3-aminopropyl)diethoxymethylsilane
Synonyms: (3-aminopropyl)diethoxymethylsilane
Molecular Weight (g/mol): 191.35
Molecular Formula: C8H21NO2Si
InChi Key: HXLAEGYMDGUSBD-UHFFFAOYSA-N
SMILES: CCO[Si](C)(CCCN)OCC
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (3)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (5)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)