accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClH3CNH2ONHNOO

CAS RN 31796-55-1

IUPAC Name: hydrogen 2-amino-N-(4-nitrophenyl)propanamide chloride
Synonyms: hydrogen 2-amino-N-(4-nitrophenyl)propanamide chloride
Molecular Weight (g/mol): 245.66
Molecular Formula: C9H12ClN3O3
InChi Key: YEXRLSXNWLNHQR-UHFFFAOYNA-N
SMILES: [H+].[Cl-].CC(N)C(=O)NC1=CC=C(C=C1)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)