accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COONHNN

CAS RN 318497-88-0

IUPAC Name: ethyl 4-cyano-1H-pyrazole-5-carboxylate
Synonyms: ethyl 4-cyano-2H-pyrazole-3-carboxylate
Molecular Weight (g/mol): 165.15
Molecular Formula: C7H7N3O2
InChi Key: COJOZBXPRZQJLK-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(C=NN1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results