CAS RN 3185-99-7
IUPAC Name:
1-methanesulfonyl-4-methylbenzene
Synonyms:
P-methylsulfonyltoluene
Molecular Weight (g/mol):
170.23
Molecular Formula:
C8H10O2S
InChi Key:
YYDNBUBMBZRNQQ-UHFFFAOYSA-N
SMILES:
CC1=CC=C(C=C1)S(C)(=O)=O