accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOFNH2

CAS RN 319-24-4

IUPAC Name: methyl 2-amino-5-fluorobenzoate
Synonyms: methyl 2-amino-5-fluorobenzoate
Molecular Weight (g/mol): 169.16
Molecular Formula: C8H8FNO2
InChi Key: PUDDYSBKCDKATP-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(N)C=CC(F)=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results