CAS RN 319-86-8
IUPAC Name:
1,2,3,4,5,6-hexachlorocyclohexane
Synonyms:
lindane hexachlorocyclohexane 林旦 hexachlorocyclohexanes hexachlorzyklohexan hexachlorcyclohexan benzene hexachloride
Molecular Weight (g/mol):
290.81
Molecular Formula:
C6H6Cl6
InChi Key:
JLYXXMFPNIAWKQ-UHFFFAOYSA-N
SMILES:
ClC1C(Cl)C(Cl)C(Cl)C(Cl)C1Cl
1
–
1
of
1
results
1
Decachlorobiphenyl Solution (BZ No. 209), 100μg/mL in Hexane, Ultra Scientific
CAS: 319-86-8 Molecular Formula: C6H6Cl6 Molecular Weight (g/mol): 290.814 InChI Key: JLYXXMFPNIAWKQ-UHFFFAOYSA-N PubChem CID: 727 ChEBI: CHEBI:24536 IUPAC Name: 1,2,3,4,5,6-hexachlorocyclohexane SMILES: C1(C(C(C(C(C1Cl)Cl)Cl)Cl)Cl)Cl