accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrN

CAS RN 31938-07-5

IUPAC Name: 2-(3-bromophenyl)acetonitrile
Synonyms: 2-(3-bromophenyl)acetonitrile
Molecular Weight (g/mol): 196.05
Molecular Formula: C8H6BrN
InChi Key: UUZYFBXKWIQKTF-UHFFFAOYSA-N
SMILES: BrC1=CC=CC(CC#N)=C1
Boiling Point 
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)