accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COO

CAS RN 3194-17-0

IUPAC Name: 1-(furan-2-yl)pentan-1-one
Synonyms: 1-(furan-2-yl)pentan-1-one
Molecular Weight (g/mol): 152.19
Molecular Formula: C9H12O2
InChi Key: HTOZHTBIOGGHDJ-UHFFFAOYSA-N
SMILES: CCCCC(=O)C1=CC=CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results