accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3195-78-6

IUPAC Name: N-ethenyl-N-methylacetamide
Synonyms: acetamide, N-ethenyl-N-methyl-
Molecular Weight (g/mol): 99.13
Molecular Formula: C5H9NO
InChi Key: PNLUGRYDUHRLOF-UHFFFAOYSA-N
SMILES: CN(C=C)C(C)=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results