accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3196-15-4

IUPAC Name: methyl 2-bromobutanoate
Synonyms: methyl 2-bromobutanoate
Molecular Weight (g/mol): 181.03
Molecular Formula: C5H9BrO2
InChi Key: UFQQDNMQADCHGH-UHFFFAOYNA-N
SMILES: CCC(Br)C(=O)OC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results