accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOH2NHCl

CAS RN 3196-73-4

IUPAC Name: methyl 3-aminopropanoate hydrochloride
Synonyms: methyl 3-aminopropanoate hydrochloride
Molecular Weight (g/mol): 139.58
Molecular Formula: C4H10ClNO2
InChi Key: XPGRZDJXVKFLHQ-UHFFFAOYSA-N
SMILES: Cl.COC(=O)CCN
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (3)
  • (3)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (3)
  • (3)
  • (2)