accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 31968-33-9

IUPAC Name: 1-(3-aminothiophen-2-yl)ethan-1-one
Synonyms: 1-(3-aminothiophen-2-yl)ethanone
Molecular Weight (g/mol): 141.19
Molecular Formula: C6H7NOS
InChi Key: MMLSKDGCLMDHAJ-UHFFFAOYSA-N
SMILES: CC(=O)C1=C(N)C=CS1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results