accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3197-06-6

IUPAC Name: 2-[(2-aminoethyl)(2-hydroxyethyl)amino]ethan-1-ol
Synonyms: 2-[(2-aminoethyl)(2-hydroxyethyl)amino]ethanol
Molecular Weight (g/mol): 148.21
Molecular Formula: C6H16N2O2
InChi Key: CYOIAXUAIXVWMU-UHFFFAOYSA-N
SMILES: NCCN(CCO)CCO
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)