accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 320-37-6

IUPAC Name: 4-nitro-2-(trifluoromethyl)benzoic acid
Synonyms: 4-nitro-2-(trifluoromethyl)benzoic acid
Molecular Weight (g/mol): 235.12
Molecular Formula: C8H4F3NO4
InChi Key: BPCKZQCTLCTDST-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(C=C(C=C1)[N+]([O-])=O)C(F)(F)F
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)