accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHFFFN

CAS RN 320-42-3

IUPAC Name: 4-hydroxy-2-(trifluoromethyl)benzonitrile
Synonyms: 4-hydroxy-2-(trifluoromethyl)benzonitrile
Molecular Weight (g/mol): 187.12
Molecular Formula: C8H4F3NO
InChi Key: MDBVIOUGHSKRMT-UHFFFAOYSA-N
SMILES: OC1=CC(=C(C=C1)C#N)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results