accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 320-60-5

IUPAC Name: 2,4-dichloro-1-(trifluoromethyl)benzene
Synonyms: 2,4-dichloro-1-(trifluoromethyl)benzene
Molecular Weight (g/mol): 215
Molecular Formula: C7H3Cl2F3
InChi Key: KALSHRGEFLVFHE-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=C(Cl)C=C1Cl
Boiling Point 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)