CAS RN 320-97-8
IUPAC Name:
4-fluorobenzene-1,2-dicarboxylic acid
Synonyms:
4-fluorobenzene-1,2-dicarboxylic acid
Molecular Weight (g/mol):
184.12
Molecular Formula:
C8H5FO4
InChi Key:
OMCXTFVBNCFZMY-UHFFFAOYSA-N
SMILES:
OC(=O)C1=CC=C(F)C=C1C(O)=O
1
–
1
of
1
results
1
Promotions Available
4-Fluorophthalic acid, 98%
4-Fluorophthalic acid, 98%
CAS: 320-97-8 Molecular Formula: C8H5FO4 Molecular Weight (g/mol): 184.12 MDL Number: MFCD06208213 InChI Key: OMCXTFVBNCFZMY-UHFFFAOYSA-N Synonym: 4-fluorobenzene-1,2-dicarboxylic acid,1,2-benzenedicarboxylic acid, 4-fluoro,5-fluorophtalic acid,5-fluorophthalic acid,4-fluorobenzene-1,2-dioic acid,3-fluorophthalicacid,pubchem1406,rarechem al bo 0829,1,2-benzenedicarboxylicacid, 4-fluoro PubChem CID: 15312 IUPAC Name: 4-fluorophthalic acid SMILES: OC(=O)C1=CC=C(F)C=C1C(O)=O