accessibility menu, dialog, popup

UserName

Viewing profile as user:
HFN

CAS RN 32001-55-1

IUPAC Name: hydrogen pyridine fluoride
Synonyms: pyridine, hydrogen fluoride
Molecular Weight (g/mol): 99.11
Molecular Formula: C5H6FN
InChi Key: GRJJQCWNZGRKAU-UHFFFAOYSA-N
SMILES: [H+].[F-].C1=CC=NC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results