accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3204-68-0

IUPAC Name: N-benzyl-N,N-dimethylanilinium chloride
Synonyms: N-benzyl-N,N-dimethylanilinium chloride
Molecular Weight (g/mol): 247.77
Molecular Formula: C15H18ClN
InChi Key: QLRKASHXFNIPLZ-UHFFFAOYSA-M
SMILES: [Cl-].C[N+](C)(CC1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results