accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOSCH3N

CAS RN 32043-95-1

IUPAC Name: ethyl 2-methyl-4-phenyl-1,3-thiazole-5-carboxylate
Synonyms: ethyl 2-methyl-4-phenyl-1,3-thiazole-5-carboxylate
Molecular Weight (g/mol): 247.31
Molecular Formula: C13H13NO2S
InChi Key: UHLMXNFHHFDVPW-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(N=C(C)S1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results