accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CO

CAS RN 3208-16-0

IUPAC Name: 2-ethylfuran
Synonyms: 2-ethylfuran
Molecular Weight (g/mol): 96.13
Molecular Formula: C6H8O
InChi Key: HLPIHRDZBHXTFJ-UHFFFAOYSA-N
SMILES: CCC1=CC=CO1
Percent Purity 
  • (3)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)