accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 32084-59-6

IUPAC Name: 6-bromo-1,4-dihydroquinazolin-4-one
Synonyms: 6-bromo-1H-quinazolin-4-one
Molecular Weight (g/mol): 225.05
Molecular Formula: C8H5BrN2O
InChi Key: OVEISJPVPHWEHR-UHFFFAOYSA-N
SMILES: BrC1=CC=C2NC=NC(=O)C2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results