accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3209-71-0

IUPAC Name: 1,2-oxazole-3-carboxylic acid
Synonyms: 3-isoxazolecarboxylic acid
Molecular Weight (g/mol): 113.07
Molecular Formula: C4H3NO3
InChi Key: UXYRXGFUANQKTA-UHFFFAOYSA-N
SMILES: OC(=O)C1=NOC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results