accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 32093-39-3

IUPAC Name: bis([bis(dimethylamino)alumanyl]dimethylamine)
Synonyms: tris(dimethylamino)aluminium dimer
Molecular Weight (g/mol): 318.43
Molecular Formula: C12H36Al2N6
InChi Key: JGZUJELGSMSOID-UHFFFAOYSA-N
SMILES: CN(C)[Al](N(C)C)N(C)C.CN(C)[Al](N(C)C)N(C)C
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)