accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3FHOO

CAS RN 321-12-0

IUPAC Name: 2-fluoro-5-methylbenzoic acid
Synonyms: 2-fluoro-5-methylbenzoic acid
Molecular Weight (g/mol): 154.14
Molecular Formula: C8H7FO2
InChi Key: UREMNBHWTNQTMS-UHFFFAOYSA-N
SMILES: CC1=CC=C(F)C(=C1)C(O)=O
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results