accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 321-28-8

IUPAC Name: 1-fluoro-2-methoxybenzene
Synonyms: 2-fluoroanisole
Molecular Weight (g/mol): 126.13
Molecular Formula: C7H7FO
InChi Key: JIXDOBAQOWOUPA-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results