accessibility menu, dialog, popup

UserName

Viewing profile as user:
F

CAS RN 321-60-8

IUPAC Name: 2-fluoro-1,1'-biphenyl
Synonyms: 1,1'-biphenyl, 2-fluoro-
Molecular Weight (g/mol): 172.2
Molecular Formula: C12H9F
InChi Key: KLECYOQFQXJYBC-UHFFFAOYSA-N
SMILES: FC1=CC=CC=C1C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (6)
Boiling Point 
  • (5)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (6)
  • (5)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)