accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 32122-09-1

IUPAC Name: ethyl 2-(benzyloxy)acetate
Synonyms: ethyl 2-(benzyloxy)acetate
Molecular Weight (g/mol): 194.23
Molecular Formula: C11H14O3
InChi Key: PHSWWKXTGAJPCQ-UHFFFAOYSA-N
SMILES: CCOC(=O)COCC1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results