accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NHNCH3OOH(?)

CAS RN 321309-43-7

IUPAC Name: 2-(3,5-dimethyl-1H-pyrazol-4-yl)benzoic acid
Synonyms: 2-(3,5-dimethyl-1H-pyrazol-4-yl)benzoic acid
Molecular Weight (g/mol): 216.24
Molecular Formula: C12H12N2O2
InChi Key: JNHKHFADRGJMJS-UHFFFAOYSA-N
SMILES: CC1=C(C(C)=NN1)C1=CC=CC=C1C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results