accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSNBOHHO

CAS RN 321438-86-2

IUPAC Name: [6-(methylsulfanyl)pyridin-3-yl]boronic acid
Synonyms: 6-(methylsulfanyl)pyridin-3-ylboronic acid
Molecular Weight (g/mol): 169.01
Molecular Formula: C6H8BNO2S
InChi Key: UXWKVPJOPVIIRU-UHFFFAOYSA-N
SMILES: CSC1=CC=C(C=N1)B(O)O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)