accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 32161-30-1

IUPAC Name: 2-amino-3-(3,4-dimethoxyphenyl)propanoic acid
Synonyms: 2-amino-3-(3,4-dimethoxyphenyl)propanoic acid
Molecular Weight (g/mol): 225.24
Molecular Formula: C11H15NO4
InChi Key: VWTFNYVAFGYEKI-UHFFFAOYNA-N
SMILES: COC1=C(OC)C=C(CC(N)C(O)=O)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)