CAS RN 321673-30-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ HDL Receptor SR-BI Inhibitor, BLT-1
The HDL Receptor SR-BI Inhibitor, BLT-1, also referenced under CAS 321673-30-7, controls the biological activity of HDL Receptor SR-B. This small molecule/inhibitor is primarily used for Phosphorylation and Dephosphorylation applications.