accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClS

CAS RN 3221-15-6

IUPAC Name: 2-(chloromethyl)thiirane
Synonyms: propane, 1-chloro-2,3-epithio-
Molecular Weight (g/mol): 108.58
Molecular Formula: C3H5ClS
InChi Key: XRWMHJJHPQTTLQ-UHFFFAOYNA-N
SMILES: ClCC1CS1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)