accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NOOH

CAS RN 3222-56-8

IUPAC Name: 2-methylpyridine-3-carboxylic acid
Synonyms: 2-methylpyridine-3-carboxylic acid
Molecular Weight (g/mol): 137.14
Molecular Formula: C7H7NO2
InChi Key: HNTZKNJGAFJMHQ-UHFFFAOYSA-N
SMILES: CC1=C(C=CC=N1)C(O)=O
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)