accessibility menu, dialog, popup

UserName

Viewing profile as user:
HN

CAS RN 32228-99-2

IUPAC Name: N-phenyl-[1,1'-biphenyl]-4-amine
Synonyms: N-phenyl-[1,1'-biphenyl]-4-amine
Molecular Weight (g/mol): 245.33
Molecular Formula: C18H15N
InChi Key: YGNUPJXMDOFFDO-UHFFFAOYSA-N
SMILES: N(C1=CC=CC=C1)C1=CC=C(C=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results