accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3226-65-1

IUPAC Name: (2S)-2-amino-4-methanesulfinylbutanoic acid
Synonyms: L-methionine sulfoxide
Molecular Weight (g/mol): 165.21
Molecular Formula: C5H11NO3S
InChi Key: QEFRNWWLZKMPFJ-YGVKFDHGSA-N
SMILES: CS(=O)CC[C@H](N)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)