accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 32279-04-2

H3COSOOOH3CSCH3OH

CAS RN 32279-04-2

IUPAC Name: (4-hydroxyphenyl)dimethylsulfanium methyl sulfate
Synonyms: (4-hydroxyphenyl)dimethylsulfanium methyl sulfate(1-)
Molecular Weight (g/mol): 266.33
Molecular Formula: C9H14O5S2
InChi Key: PKQZVASULXKBJV-UHFFFAOYSA-N
SMILES: COS([O-])(=O)=O.C[S+](C)C1=CC=C(O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results