CAS RN 3228-02-2
IUPAC Name:
3-methyl-4-(propan-2-yl)phenol
Synonyms:
o-cymen-5-ol p-thymol frecide o-cymen-5y-ol biosal (antibacterial) isopropyl cresols
Molecular Weight (g/mol):
150.22
Molecular Formula:
C10H14O
InChi Key:
IJALWSVNUBBQRA-UHFFFAOYSA-N
SMILES:
CC(C)C1=CC=C(O)C=C1C
1
–
1
of
1
results
1
4-Isopropyl-3-methylphenol 99.0+%, TCI America™
CAS: 3228-02-2 Molecular Formula: C10H14O Molecular Weight (g/mol): 150.22 MDL Number: MFCD00010704 InChI Key: IJALWSVNUBBQRA-UHFFFAOYSA-N Synonym: Biosol, 4-Isopropyl-m-cresol PubChem CID: 18597 ChEBI: CHEBI:34429 IUPAC Name: 3-methyl-4-propan-2-ylphenol SMILES: CC1=C(C=CC(=C1)O)C(C)C