accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 32281-97-3

IUPAC Name: 7-bromo-1,2,3,4-tetrahydronaphthalen-1-one
Synonyms: 7-bromo-3,4-dihydro-2H-naphthalen-1-one
Molecular Weight (g/mol): 225.09
Molecular Formula: C10H9BrO
InChi Key: YGVDCGFUUUJCDF-UHFFFAOYSA-N
SMILES: BrC1=CC=C2CCCC(=O)C2=C1
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (3)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (4)
  • (2)
  • (4)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)