accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHCH2HC

CAS RN 3230-69-1

IUPAC Name: 3-methylpent-1-en-4-yn-3-ol
Synonyms: 1-penten-4-yn-3-ol, 3-methyl-
Molecular Weight (g/mol): 96.13
Molecular Formula: C6H8O
InChi Key: VBATUBQIYXCZPA-UHFFFAOYNA-N
SMILES: CC(O)(C=C)C#C
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results