accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 32314-39-9

IUPAC Name: 4-chloro-2,8-dimethylquinoline
Synonyms: 4-chloro-2,8-dimethylquinoline
Molecular Weight (g/mol): 191.66
Molecular Formula: C11H10ClN
InChi Key: ILOAZUIOTZZIBK-UHFFFAOYSA-N
SMILES: CC1=CC(Cl)=C2C=CC=C(C)C2=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results